siRNA Details
| Gene Symbol: SCRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007281.2 | NP_009212.1 | 2 | 612 | ||
| XM_011531578.1 | XP_011529880.1 | 3 | 331 | |||
| XM_017007678.1 | XP_016863167.1 | 3 | 325 | |||
| GenBank mRNA | AJ224677.1 | - | 2 | 612 | ||
| AK312146.1 | - | 2 | 261 | |||
| AY359040.1 | - | 2 | 359 | |||
| BC017583.2 | - | 2 | 391 | |||
| CR542142.1 | - | 1 | 199 | |||

SCRG1
Genomic Map