siRNA Details
| Gene Symbol: GLUD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012084.3 | NP_036216.2 | 1 | 1997 | ||
| GenBank mRNA | BC005111.1 | - | 1 | 1040 | ||
| BC050732.2 | - | 1 | 1997 | 6164919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLUD2| Gene Symbol: GLUD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012084.3 | NP_036216.2 | 1 | 1997 | ||
| GenBank mRNA | BC005111.1 | - | 1 | 1040 | ||
| BC050732.2 | - | 1 | 1997 | 6164919 | ||