siRNA Details
| Gene Symbol: GPR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098200.1 | NP_001091670.1 | 2 | 878 | ||
| NM_005292.3 | NP_005283.1 | 3 | 1070 | |||
| XM_006719946.3 | XP_006720009.1 | 3 | 1880 | |||
| GenBank mRNA | AF261135.1 | - | 2 | 753 | ||
| AF339781.1 | - | 2 | 826 | |||
| BC008569.1 | - | 2 | 837 | 4338810 | ||
| BC050646.1 | - | 3 | 1016 | 4341573 | ||
| BC066927.1 | - | 3 | 1126 | 4373724 | ||
| BT009907.1 | - | 1 | 569 | |||
| JQ308176.1 | - | 1 | 569 | |||

GPR18
Genomic Map