siRNA Details
Gene Symbol: GPR27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018971.1 | NP_061844.1 | 1 | 386 | ||
GenBank mRNA | AB040799.1 | - | 1 | 386 | ||
HQ709187.1 | - | 1 | 386 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
