siRNA Details
| Gene Symbol: HOXD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002148.3 | NP_002139.2 | 1 | 285 | ||
| GenBank mRNA | AK309451.1 | - | 1 | 269 | ||
| AL526269.3 | - | 1 | 285 | |||
| BC069619.1 | - | 1 | 265 | 7262455 | ||
| BC074760.2 | - | 1 | 264 | 30915219 | ||
| X59373.1 | - | 1 | 248 | |||

HOXD10
Genomic Map