siRNA Details
| Gene Symbol: AGFG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006076.4 | NP_006067.3 | 11, 12 | 1524 | ||
| XM_005250306.2 | XP_005250363.1 | 12, 13 | 1543 | |||
| XM_017012112.1 | XP_016867601.1 | 11, 12 | 1465 | |||
| XM_017012114.1 | XP_016867603.1 | 10, 11 | 1223 | |||
| GenBank mRNA | AF015042.1 | - | 11, 12 | 1448 | ||
| AK314954.1 | - | 11, 12 | 1510 | |||
| BC017329.2 | - | 11, 12 | 1476 | 4810320 | ||

AGFG2
Genomic Map