siRNA Details
| Gene Symbol: ENPP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178543.4 | NP_848638.3 | 3 | 757 | ||
| XM_011524737.1 | XP_011523039.1 | 3 | 941 | |||
| GenBank mRNA | AK126250.1 | - | 3 | 941 | ||
| AY230663.1 | - | 3 | 740 | |||
| AY358622.1 | - | 3 | 723 | |||
| BC041453.1 | - | 3 | 740 | 5186743 | ||

ENPP7
Genomic Map