siRNA Details
| Gene Symbol: INHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002191.3 | NP_002182.1 | 2 | 424 | ||
| GenBank mRNA | BC006391.2 | - | 2 | 361 | 4126990 | |
| BT006954.1 | - | 2 | 281 | |||
| M13144.1 | - | 2 | 238 | |||
| M13981.1 | - | 2 | 425 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INHA| Gene Symbol: INHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002191.3 | NP_002182.1 | 2 | 424 | ||
| GenBank mRNA | BC006391.2 | - | 2 | 361 | 4126990 | |
| BT006954.1 | - | 2 | 281 | |||
| M13144.1 | - | 2 | 238 | |||
| M13981.1 | - | 2 | 425 | |||