siRNA Details
| Gene Symbol: INHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002191.3 | NP_002182.1 | 2 | 1195 | ||
| GenBank mRNA | BC006391.2 | - | 2 | 1132 | 4126990 | |
| BT006954.1 | - | 2 | 1052 | |||
| M13144.1 | - | 2 | 1009 | |||
| M13981.1 | - | 2 | 1196 | |||

INHA
Genomic Map