siRNA Details
| Gene Symbol: ITGAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002208.4 | NP_002199.3 | 21 | 2658 | ||
| XM_011523823.2 | XP_011522125.1 | 21 | 3096 | |||
| XM_011523825.2 | XP_011522127.1 | 21 | 3094 | |||
| XM_011523827.2 | XP_011522129.1 | 21 | 3088 | |||
| XM_011523828.2 | XP_011522130.1 | 21 | 3092 | |||
| XM_017024586.1 | XP_016880075.1 | 22 | 4481 | |||
| XM_017024587.1 | XP_016880076.1 | 21 | 4407 | |||
| GenBank mRNA | BC113436.1 | - | 21 | 2648 | 8322488 | |
| BC117207.1 | - | 21 | 2648 | 40125758 | ||
| L25851.2 | - | 21 | 2684 | |||

ITGAE
Genomic Map