siRNA Details
| Gene Symbol: KIF5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004521.2 | NP_004512.1 | 3, 4 | 759 | ||
| XM_017016224.1 | XP_016871713.1 | 3, 4 | 365 | |||
| GenBank mRNA | AK289413.1 | - | 3, 4 | 705 | ||
| AK307502.1 | - | 4, 5 | 456 | |||
| BC010885.1 | - | 3, 4 | 695 | |||
| BC065267.1 | - | 3, 4 | 716 | |||
| BC080604.1 | - | 3, 4 | 690 | |||
| BC111042.1 | - | 3, 4 | 699 | |||
| BC126279.1 | - | 3, 4 | 512 | 8991995 | ||
| BC126281.1 | - | 3, 4 | 512 | 8991997 | ||
| FJ224324.1 | - | 3, 4 | 602 | |||
| X65873.1 | - | 3, 4 | 602 | |||

KIF5B
Genomic Map