siRNA Details
| Gene Symbol: NDUFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004542.3 | NP_004533.1 | 2, 3 | 115 | ||
| XM_017026833.1 | XP_016882322.1 | 3, 4 | 532 | |||
| GenBank mRNA | AF044955.1 | - | 2, 3 | 107 | ||
| AF070653.1 | - | 2, 3 | 116 | |||
| AK289489.1 | - | 2, 3 | 113 | |||
| BC022369.1 | - | 2, 3 | 112 | 4737656 | ||
| BC061644.1 | - | 2, 3 | 115 | 5748578 | ||
| CR542143.1 | - | 1, 2 | 88 | |||

NDUFA3
Genomic Map