siRNA Details
| Gene Symbol: XRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317960.1 | NP_001304889.1 | 9 | 1016 | ||
| NM_012255.4 | NP_036387.2 | 9 | 856 | |||
| XM_017027722.1 | XP_016883211.1 | 9 | 1011 | |||
| XM_017027723.1 | XP_016883212.1 | 9 | 785 | |||
| XM_017027724.1 | XP_016883213.1 | 9 | 1011 | |||
| GenBank mRNA | AF064257.1 | - | 9 | 809 | ||
| AK001573.1 | - | 9 | 826 | |||
| AK302846.1 | - | 9 | 720 | |||
| AK303312.1 | - | 8 | 667 | |||
| AK314602.1 | - | 9 | 795 | |||
| AL136841.1 | - | 9 | 782 | |||
| AY382900.1 | - | 8 | 671 | |||

XRN2
Genomic Map