siRNA Details
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024940.2 | NP_001020111.1 | 7 | 1977 | ||
| NM_016102.3 | NP_057186.1 | 7 | 2013 | |||
| XM_006711779.2 | XP_006711842.1 | 7 | 2040 | |||
| XM_011544209.2 | XP_011542511.1 | 7 | 2027 | |||
| XM_011544210.2 | XP_011542512.1 | 7 | 2129 | |||
| XM_011544211.2 | XP_011542513.1 | 7 | 2030 | |||
| XM_017001418.1 | XP_016856907.1 | 5 | 3001 | |||
| XM_017001419.1 | XP_016856908.1 | 6 | 2545 | |||
| GenBank mRNA | AK055426.1 | - | 6 | 2581 | ||

TRIM17
Genomic Map