siRNA Details
| Gene Symbol: FKBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143782.1 | NP_001137254.1 | 2 | 297 | ||
| NM_016594.2 | NP_057678.1 | 2 | 297 | |||
| GenBank mRNA | AF238079.1 | - | 2 | 250 | ||
| AK301455.1 | - | 2 | 270 | |||
| AY358998.1 | - | 2 | 255 | |||
| BC027973.1 | - | 2 | 249 | 5229643 | ||
| BC048292.1 | - | 2 | 260 | 5207367 | ||
| DA378205.1 | - | 2 | 297 | |||

FKBP11
Genomic Map