siRNA Details
| Gene Symbol: HEATR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018072.5 | NP_060542.4 | 23 | 3392 | ||
| GenBank mRNA | AK001857.1 | - | 1 | 662 | ||
| AK304129.1 | - | 3 | 469 | |||
| AK316413.1 | - | 3 | 363 | |||
| BC065205.1 | - | 1 | 128 | |||
| BC130000.1 | - | 8 | 1316 | |||
| BC150614.1 | - | 22 | 3262 | 9056984 | ||

HEATR1
Genomic Map