siRNA Details
| Gene Symbol: THAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018105.2 | NP_060575.1 | 3 | 1246 | ||
| NM_199003.1 | NP_945354.1 | 2 | 1050 | |||
| GenBank mRNA | AK001339.1 | - | 3 | 1242 | ||
| AL832077.1 | - | 2 | 1024 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP1| Gene Symbol: THAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018105.2 | NP_060575.1 | 3 | 1246 | ||
| NM_199003.1 | NP_945354.1 | 2 | 1050 | |||
| GenBank mRNA | AK001339.1 | - | 3 | 1242 | ||
| AL832077.1 | - | 2 | 1024 | |||