siRNA Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297760.1 | NP_001284689.1 | 3 | 938 | ||
| NM_001297761.1 | NP_001284690.1 | 2 | 678 | |||
| NM_018208.3 | NP_060678.2 | 3 | 938 | |||
| XM_011509714.2 | XP_011508016.1 | 2 | 178 | |||
| XM_011509715.1 | XP_011508017.1 | 2 | 483 | |||
| XM_017001632.1 | XP_016857121.1 | 2 | 266 | |||
| GenBank mRNA | AB073608.1 | - | 3 | 538 | ||
| AK001623.1 | - | 3 | 650 | |||
| AK293426.1 | - | 2 | 386 | |||
| AK316493.1 | - | 2 | 174 | |||
| BC010082.2 | - | 3 | 759 | 4329734 | ||

ETNK2
Genomic Map