siRNA Details
| Gene Symbol: SIRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039508.1 | NP_001034597.1 | 5 | 1222 | ||
| NM_018556.3 | NP_061026.2 | 6 | 1555 | |||
| NM_080816.2 | NP_543006.2 | 4 | 904 | |||
| XM_005260749.3 | XP_005260806.1 | 5 | 1273 | |||
| XM_011529286.2 | XP_011527588.1 | 6 | 1602 | |||
| GenBank mRNA | AB042624.1 | - | 6 | 1571 | ||
| AK124224.1 | - | 1 | 3935 | |||
| AK292745.1 | - | 6 | 1555 | |||
| BC020629.1 | - | 4 | 881 | |||
| BC064532.1 | - | 6 | 1516 | 5588038 | ||

SIRPG
Genomic Map