siRNA Details
| Gene Symbol: TRIM36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300752.1 | NP_001287681.1 | 7 | 1559 | ||
| NM_001300759.1 | NP_001287688.1 | 8 | 1578 | |||
| NM_018700.3 | NP_061170.2 | 8 | 1794 | |||
| XM_017009621.1 | XP_016865110.1 | 10 | 1806 | |||
| XM_017009622.1 | XP_016865111.1 | 10 | 2523 | |||
| XM_017009623.1 | XP_016865112.1 | 10 | 1806 | |||
| GenBank mRNA | AB196558.1 | - | 7 | 1050 | ||
| AJ272269.1 | - | 8 | 1411 | |||
| AK058179.1 | - | 3 | 435 | |||
| AK296389.1 | - | 8 | 1576 | |||
| AK316422.1 | - | 8 | 1481 | |||

TRIM36
Genomic Map