siRNA Details
| Gene Symbol: PLXNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017514.4 | NP_059984.3 | 32 | 5668 | ||
| XM_005274705.4 | XP_005274762.2 | 32 | 5608 | |||
| XM_005274706.4 | XP_005274763.2 | 32 | 5340 | |||
| XM_006724829.3 | XP_006724892.1 | 32 | 5572 | |||
| XM_011531183.2 | XP_011529485.1 | 31 | 5032 | |||
| GenBank mRNA | X87852.1 | - | 32 | 5677 | ||

PLXNA3
Genomic Map