siRNA Details
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 9 | 1600 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: METTL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018396.2 | NP_060866.2 | 9 | 1600 |