siRNA Details
| Gene Symbol: UBL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048241.2 | NP_001041706.1 | 5 | 424 | ||
| NM_024292.3 | NP_077268.1 | 5 | 492 | |||
| GenBank mRNA | AF313915.1 | - | 5 | 373 | ||
| AK128885.1 | - | 5 | 492 | |||
| BC110839.1 | - | 5 | 340 | 6474495 | ||
| BP213482.1 | - | 5 | 412 | |||

UBL5
Genomic Map