siRNA Details
| Gene Symbol: RGS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005339.1 | NP_001005339.1 | 2 | 226 | ||
| NM_002925.3 | NP_002916.1 | 2 | 175 | |||
| XM_017016492.1 | XP_016871981.1 | 2 | 223 | |||
| XM_017016493.1 | XP_016871982.1 | 2 | 268 | |||
| GenBank mRNA | AF045229.2 | - | 2 | 155 | ||
| AF368902.1 | - | 1 | 130 | |||
| AF493934.1 | - | 1 | 112 | |||
| AK290773.1 | - | 2 | 181 | |||
| AL547312.3 | - | 2 | 175 | |||
| AV707839.1 | - | 2 | 175 | |||
| BC009361.2 | - | 2 | 171 | 4301597 | ||
| BF685740.1 | - | 2 | 172 | |||
| BG763741.1 | - | 2 | 182 | |||
| BM914660.1 | - | 225 | ||||
| CB124184.1 | - | 2 | 226 | |||
| CR457008.1 | - | 1 | 112 | |||

RGS10
Genomic Map