siRNA Details
| Gene Symbol: CEP162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286206.1 | NP_001273135.1 | 7 | 676 | ||
| NM_014895.3 | NP_055710.2 | 7 | 730 | |||
| XM_005248674.3 | XP_005248731.1 | 6 | 853 | |||
| XM_005248678.3 | XP_005248735.1 | 7 | 984 | |||
| XM_006715380.2 | XP_006715443.1 | 8 | 824 | |||
| XM_011535594.2 | XP_011533896.1 | 7 | 984 | |||
| XM_017010483.1 | XP_016865972.1 | 6 | 1409 | |||
| XM_017010484.1 | XP_016865973.1 | 8 | 1027 | |||
| GenBank mRNA | AB023226.1 | - | 7 | 637 | ||
| AK308190.1 | - | 7 | 697 | |||
| BC063632.1 | - | 7 | 703 | |||

CEP162
Genomic Map