siRNA Details
| Gene Symbol: CCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002981.2 | NP_002972.1 | 3 | 429 | ||
| GenBank mRNA | DA620119.1 | - | 3 | 427 | ||
| M57502.1 | - | 3 | 407 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCL1| Gene Symbol: CCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002981.2 | NP_002972.1 | 3 | 429 | ||
| GenBank mRNA | DA620119.1 | - | 3 | 427 | ||
| M57502.1 | - | 3 | 407 | |||