siRNA Details
| Gene Symbol: RRAGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022157.3 | NP_071440.1 | 2 | 472 | ||
| GenBank mRNA | AF272035.1 | - | 2 | 300 | ||
| AF323609.1 | - | 2 | 295 | |||
| AK315791.1 | - | 2 | 397 | |||
| BC016668.1 | - | 2 | 317 | 4342448 | ||
| BG717489.1 | - | 2 | 480 | |||
| BI461665.1 | - | 2 | 422 | |||

RRAGC
Genomic Map