siRNA Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145365.1 | NP_001138837.1 | 6 | 2206 | ||
| NM_014897.2 | NP_055712.1 | 6 | 2061 | |||
| NR_135579.1 | 7 | 1790 | ||||
| XM_005257166.2 | XP_005257223.1 | 5 | 1413 | |||
| GenBank mRNA | AB023141.1 | - | 6 | 2061 | ||
| BC139779.1 | - | 5 | 1916 | 40122232 | ||
| BC152440.1 | - | 6 | 2061 | 8862563 | ||
| CR933680.1 | - | 6 | 2057 | |||

ZNF652
Genomic Map