siRNA Details
| Gene Symbol: EDDM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022360.4 | NP_071755.1 | 2 | 394 | ||
| GenBank mRNA | AY358865.1 | - | 2 | 394 | ||
| BC128030.1 | - | 2 | 357 | 40002822 | ||
| BX112699.1 | - | 2 | 390 | |||
| X76386.2 | - | 1 | 356 | |||

EDDM3B
Genomic Map