siRNA Details
| Gene Symbol: EDDM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022360.4 | NP_071755.1 | 2 | 489 | ||
| GenBank mRNA | AY358865.1 | - | 2 | 489 | ||
| BC128030.1 | - | 2 | 452 | 40002822 | ||
| X76386.2 | - | 1 | 451 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDDM3B| Gene Symbol: EDDM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022360.4 | NP_071755.1 | 2 | 489 | ||
| GenBank mRNA | AY358865.1 | - | 2 | 489 | ||
| BC128030.1 | - | 2 | 452 | 40002822 | ||
| X76386.2 | - | 1 | 451 | |||