siRNA Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320666.1 | NP_001307595.1 | 6 | 1374 | ||
| NM_001320667.1 | NP_001307596.1 | 6 | 1280 | |||
| NM_001320668.1 | NP_001307597.1 | 6 | 1150 | |||
| NM_001320669.1 | NP_001307598.1 | 6 | 1088 | |||
| NM_001320670.1 | NP_001307599.1 | 6 | 1085 | |||
| NM_001320671.1 | NP_001307600.1 | 6 | 991 | |||
| NM_014898.2 | NP_055713.1 | 6 | 1321 | |||
| GenBank mRNA | AB023178.1 | - | 6 | 1321 | ||
| BC041087.1 | - | 6 | 1350 | 5579247 | ||
| BC050542.1 | - | 6 | 1279 | |||
| GU388430.1 | - | 6 | 1150 | |||

ZFP30
Genomic Map