siRNA Details
| Gene Symbol: ZNF747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305018.1 | NP_001291947.1 | 2 | 614 | ||
| NM_001305019.1 | NP_001291948.1 | 2 | 614 | |||
| NM_001305020.1 | NP_001291949.1 | 1 | 754 | |||
| NM_023931.3 | NP_076420.1 | 1 | 754 | |||
| GenBank mRNA | AB209546.1 | - | 1 | 627 | ||
| AK054567.1 | - | 1 | 656 | |||
| AK292192.1 | - | 1 | 465 | |||
| AK299308.1 | - | 2 | 499 | |||
| AK316254.1 | - | 2 | 542 | |||
| BC001361.2 | - | 1 | 657 | 3050694 | ||
| DC305764.1 | - | 2 | 502 | |||

ZNF747
Genomic Map