siRNA Details
| Gene Symbol: SRPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003137.4 | NP_003128.3 | 11 | 1199 | ||
| NR_034069.1 | 11 | 1227 | ||||
| GenBank mRNA | AJ318054.1 | - | 10 | 1597 | ||
| AK292277.1 | - | 11 | 1144 | |||
| AK299591.1 | - | 11 | 1187 | |||
| AK312712.1 | - | 11 | 1091 | |||
| BC038292.1 | - | 11 | 1164 | 4824261 | ||
| BX648663.1 | - | 11 | 1116 | |||
| BX648697.1 | - | 11 | 1117 | |||

SRPK1
Genomic Map