siRNA Details
| Gene Symbol: SRPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003137.4 | NP_003128.3 | 7 | 617 | ||
| NR_034069.1 | 7 | 645 | ||||
| GenBank mRNA | AJ318054.1 | - | 6 | 1015 | ||
| AK292277.1 | - | 7 | 562 | |||
| AK299591.1 | - | 7 | 605 | |||
| AK312712.1 | - | 7 | 509 | |||
| BC038292.1 | - | 7 | 582 | 4824261 | ||
| BG772464.1 | - | 7 | 625 | |||
| BX648663.1 | - | 7 | 534 | |||
| BX648697.1 | - | 7 | 535 | |||

SRPK1
Genomic Map