siRNA Details
| Gene Symbol: SSX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278691.1 | NP_001265620.1 | 4 | 305 | ||
| NM_005635.3 | NP_005626.1 | 3 | 219 | |||
| GenBank mRNA | BC001003.2 | - | 3 | 164 | 3445470 | |
| BC125151.1 | - | 3 | 162 | 40120554 | ||
| BC128611.1 | - | 3 | 161 | 6159943 | ||
| BC133693.1 | - | 3 | 162 | 40120550 | ||
| BC150487.2 | - | 3 | 166 | 40120555 | ||
| BE777476.1 | - | 4 | 240 | |||
| BI831262.1 | - | 3 | 217 | |||
| X86174.1 | - | 3 | 174 | |||

SSX1
Genomic Map