siRNA Details
| Gene Symbol: TESK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318230.1 | NP_001305159.1 | 8, 9 | 1257 | ||
| NM_006285.2 | NP_006276.2 | 9, 10 | 1340 | |||
| GenBank mRNA | AK298907.1 | - | 9, 10 | 878 | ||
| BC037561.1 | - | 9, 10 | 935 | |||
| BC038448.1 | - | 9, 10 | 1314 | 5170888 | ||
| BC067130.1 | - | 9, 10 | 1351 | 6153652 | ||
| D50863.1 | - | 9, 10 | 1276 | |||

TESK1
Genomic Map