siRNA Details
| Gene Symbol: TTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166691.1 | NP_001160163.1 | 7 | 882 | ||
| NM_003318.4 | NP_003309.2 | 7 | 870 | |||
| XM_011536099.2 | XP_011534401.1 | 7 | 844 | |||
| XM_011536100.2 | XP_011534402.1 | 7 | 870 | |||
| XM_017011242.1 | XP_016866731.1 | 8 | 1399 | |||
| GenBank mRNA | AK095599.1 | - | 7 | 1342 | ||
| AK292460.1 | - | 7 | 845 | |||
| AK315696.1 | - | 7 | 787 | |||
| AX775733.1 | - | 7 | 842 | |||
| BC000633.2 | - | 7 | 833 | 3343925 | ||
| BC032858.2 | - | 7 | 835 | 5270012 | ||

TTK
Genomic Map