siRNA Details
Gene Symbol: UBE2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003339.2 | NP_003330.1 | 2 | 704 | ||
NM_181838.1 | NP_862821.1 | 3 | 868 | |||
XM_017009820.1 | XP_016865309.1 | 5 | 1075 | |||
GenBank mRNA | AK001311.1 | - | 2 | 477 | ||
AK001428.1 | - | 2 | 531 | |||
AK310273.1 | - | 2 | 433 | |||
AY651263.1 | - | 2 | 460 | |||
BC019052.2 | - | 2 | 488 | |||
BC033349.1 | - | 2 | 704 | 4827210 | ||
BQ186099.1 | - | 3 | 353 | |||
U39317.1 | - | 2 | 100 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019912, AK204863, AK199554, AK005355, U62483, and others.
Panther Classification:
Gene Ontology Categories:
