siRNA Details
| Gene Symbol: ZNF136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003437.3 | NP_003428.1 | 4 | 1584 | ||
| XM_011528269.2 | XP_011526571.1 | 5 | 2079 | |||
| XM_017027243.1 | XP_016882732.1 | 3 | 1842 | |||
| GenBank mRNA | AK123008.1 | - | 3 | 2601 | ||
| AK291242.1 | - | 4 | 1579 | |||
| AK299601.1 | - | 3 | 1407 | |||
| AY500356.1 | - | 1 | 99 | |||
| BC006421.1 | - | 4 | 1537 | 4040430 | ||
| U09367.1 | - | 4 | 1508 | |||

ZNF136
Genomic Map