siRNA Details
| Gene Symbol: ZNF230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006300.3 | NP_006291.2 | 5 | 1262 | ||
| GenBank mRNA | BC050340.1 | - | 5 | 1239 | 4821663 | |
| BC094708.1 | - | 5 | 1262 | 30402948 | ||
| U95044.1 | - | 5 | 1231 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF230| Gene Symbol: ZNF230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006300.3 | NP_006291.2 | 5 | 1262 | ||
| GenBank mRNA | BC050340.1 | - | 5 | 1239 | 4821663 | |
| BC094708.1 | - | 5 | 1262 | 30402948 | ||
| U95044.1 | - | 5 | 1231 | |||