siRNA Details
| Gene Symbol: KREMEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253725.1 | NP_001240654.1 | 8 | 1805 | ||
| NM_001253726.1 | NP_001240655.1 | 9 | 1884 | |||
| NM_024507.3 | NP_078783.1 | 8 | 1922 | |||
| NM_172229.2 | NP_757384.1 | 9 | 2001 | |||
| GenBank mRNA | AK027669.1 | - | 8 | 1826 | ||
| AK075033.1 | - | 8 | 1746 | |||
| AK223422.1 | - | 8 | 1694 | |||
| AK301953.1 | - | 9 | 1825 | |||
| BC003533.1 | - | 8 | 1729 | 3609280 | ||
| BC009383.2 | - | 7 | 2286 | 4127918 | ||

KREMEN2
Genomic Map