siRNA Details
Gene Symbol: TFPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271003.1 | NP_001257932.1 | 4 | 867 | ||
NM_001271004.1 | NP_001257933.1 | 4 | 824 | |||
NM_006528.3 | NP_006519.1 | 4 | 900 | |||
GenBank mRNA | AK024817.1 | - | 4 | 662 | ||
AK075425.1 | - | 4 | 585 | |||
AK075427.1 | - | 4 | 712 | |||
AK092499.1 | - | 4 | 629 | |||
AK129833.1 | - | 4 | 662 | |||
AK313260.1 | - | 4 | 662 | |||
AY691946.1 | - | 4 | 638 | |||
BC005330.1 | - | 4 | 662 | 3929933 | ||
BC034155.1 | - | 3 | 385 | |||
D29992.1 | - | 4 | 643 | |||
DA833200.1 | - | 4 | 585 | |||
L27624.1 | - | 4 | 625 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
