siRNA Details
| Gene Symbol: PODNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146254.1 | NP_001139726.1 | 8 | 1067 | ||
| NM_001146255.1 | NP_001139727.1 | 6 | 800 | |||
| NM_024825.3 | NP_079101.3 | 8 | 1091 | |||
| XM_006722901.1 | XP_006722964.1 | 8 | 1024 | |||
| XM_006722902.1 | XP_006722965.1 | 7 | 889 | |||
| XM_006722903.1 | XP_006722966.1 | 6 | 757 | |||
| XM_011528308.1 | XP_011526610.1 | 8 | 1058 | |||
| XM_011528309.2 | XP_011526611.1 | 7 | 983 | |||
| XM_011528310.2 | XP_011526612.1 | 7 | 946 | |||
| XM_011528311.1 | XP_011526613.1 | 7 | 925 | |||
| XM_011528313.1 | XP_011526615.1 | 7 | 993 | |||
| XM_011528314.1 | XP_011526616.1 | 7 | 914 | |||
| XM_011528316.1 | XP_011526618.1 | 5 | 726 | |||
| XM_017027313.1 | XP_016882802.1 | 7 | 948 | |||
| GenBank mRNA | AK027100.1 | - | 6 | 791 | ||
| AK290731.1 | - | 8 | 1024 | |||
| AK296101.1 | - | 6 | 787 | |||
| AK298489.1 | - | 8 | 1067 | |||
| BC057786.1 | - | 8 | 1091 | 30347370 | ||

PODNL1
Genomic Map