siRNA Details
| Gene Symbol: DOLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014908.3 | NP_055723.1 | 1 | 1626 | ||
| GenBank mRNA | AB029017.1 | - | 1 | 1512 | ||
| AK225402.1 | - | 2 | 911 | |||
| AK300180.1 | - | 2 | 1244 | |||
| AY358759.1 | - | 1 | 1481 | |||
| BC035556.1 | - | 1 | 1479 | 5224439 | ||
| KU178553.1 | - | 1 | 1196 | |||

DOLK
Genomic Map