siRNA Details
| Gene Symbol: THAP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008695.1 | NP_001008695.1 | 5 | 1053 | ||
| NM_030573.2 | NP_085050.2 | 4 | 1227 | |||
| GenBank mRNA | AK024609.1 | - | 2 | 1794 | ||
| AK025906.1 | - | 2 | 1559 | |||
| AK300715.1 | - | 5 | 842 | |||
| BC004346.1 | - | 4 | 1198 | 3633743 | ||

THAP7
Genomic Map