siRNA Details
| Gene Symbol: GPR68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177676.1 | NP_001171147.1 | 2 | 2113 | ||
| NM_003485.3 | NP_003476.3 | 2 | 2067 | |||
| XM_005268110.4 | XP_005268167.1 | 2 | 3253 | |||
| XM_005268111.3 | XP_005268168.1 | 2 | 2033 | |||
| XM_005268112.3 | XP_005268169.1 | 2 | 1973 | |||
| XM_006720262.3 | XP_006720325.1 | 2 | 1999 | |||
| XM_011537196.2 | XP_011535498.1 | 3 | 2117 | |||
| XM_011537197.2 | XP_011535499.1 | 2 | 2397 | |||
| XM_011537198.2 | XP_011535500.1 | 2 | 2623 | |||
| XM_011537199.2 | XP_011535501.1 | 2 | 2068 | |||
| GenBank mRNA | BC098567.1 | - | 2 | 2010 | 30925589 | |
More Information
Set Membership: |
|

GPR68
Genomic Map