siRNA Details
| Gene Symbol: SOX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031439.3 | NP_113627.1 | 2 | 1843 | ||
| GenBank mRNA | AK055556.1 | - | 2 | 1781 | ||
| BC004299.1 | - | 2 | 1770 | |||
| BC071947.1 | - | 2 | 1769 | 6459570 | ||
| BU187815.1 | - | 1 | 162 | |||

SOX7
Genomic Map