siRNA Details
| Gene Symbol: RASSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032023.3 | NP_114412.2 | 6 | 545 | ||
| XM_005271833.4 | XP_005271890.1 | 6 | 595 | |||
| XM_005271835.4 | XP_005271892.1 | 6 | 545 | |||
| XM_006718013.3 | XP_006718076.1 | 6 | 640 | |||
| XM_006718014.3 | XP_006718077.1 | 5 | 2059 | |||
| XM_011540255.2 | XP_011538557.1 | 5 | 420 | |||
| XM_017016754.1 | XP_016872243.1 | 5 | 2185 | |||
| XM_017016755.1 | XP_016872244.1 | 5 | 2140 | |||
| XM_017016756.1 | XP_016872245.1 | 6 | 666 | |||
| XM_017016757.1 | XP_016872246.1 | 5 | 2211 | |||
| XM_017016758.1 | XP_016872247.1 | 4 | 1916 | |||
| GenBank mRNA | AB209446.1 | - | 5 | 2417 | ||
| AF260335.1 | - | 6 | 509 | |||
| AK055763.1 | - | 4 | 1553 | |||
| AK092770.1 | - | 3 | 1755 | |||
| AK097272.1 | - | 1 | 1823 | |||
| AY216713.1 | - | 6 | 545 | |||
| AY216714.1 | - | 6 | 505 | |||
| AY216715.1 | - | 6 | 572 | |||
| AY216716.1 | - | 5 | 453 | |||
| BC032593.1 | - | 6 | 527 | 5527647 | ||

RASSF4
Genomic Map