siRNA Details
| Gene Symbol: CREB3L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271995.1 | NP_001258924.1 | 5, 6 | 869 | ||
| NM_001271996.1 | NP_001258925.1 | 5, 6 | 866 | |||
| NM_032607.2 | NP_115996.1 | 5, 6 | 872 | |||
| GenBank mRNA | AB050902.1 | - | 5, 6 | 841 | ||
| AB073612.1 | - | 5, 6 | 830 | |||
| AK172839.1 | - | 5, 6 | 841 | |||
| AK313099.1 | - | 5, 6 | 844 | |||
| BC101504.1 | - | 5, 6 | 805 | 8069010 | ||
| BC101508.1 | - | 5, 6 | 801 | 8069014 | ||
| BC143609.1 | - | 5, 6 | 799 | 9052122 | ||

CREB3L3
Genomic Map