siRNA Details
Gene Symbol: DGKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003648.2 | NP_003639.2 | 17 | 2174 | ||
NM_152879.2 | NP_690618.2 | 18 | 2239 | |||
XM_005246107.3 | XP_005246164.1 | 16 | 1906 | |||
XM_005246108.1 | XP_005246165.1 | 16 | 2060 | |||
XM_005246110.3 | XP_005246167.1 | 7 | 1911 | |||
XM_011512028.2 | XP_011510330.1 | 18 | 3698 | |||
XM_011512029.2 | XP_011510331.1 | 18 | 3699 | |||
XM_011512030.2 | XP_011510332.1 | 18 | 2295 | |||
XM_011512031.2 | XP_011510333.1 | 18 | 2169 | |||
XM_011512032.2 | XP_011510334.1 | 18 | 2153 | |||
XM_011512033.2 | XP_011510335.1 | 18 | 3700 | |||
XM_011512035.1 | XP_011510337.1 | 16 | 2040 | |||
XM_011512037.2 | XP_011510339.1 | 16 | 2144 | |||
XM_011512039.2 | XP_011510341.1 | 18 | 3700 | |||
XM_011512040.2 | XP_011510342.1 | 16 | 2076 | |||
XM_017005138.1 | XP_016860627.1 | 18 | 2577 | |||
XM_017005139.1 | XP_016860628.1 | 16 | 2069 | |||
XM_017005140.1 | XP_016860629.1 | 18 | 2171 | |||
GenBank mRNA | AB078966.1 | - | 18 | 2239 | ||
AK307426.1 | - | 6 | 797 | |||
AK308846.1 | - | 16 | 2060 | |||
AK311044.1 | - | 16 | 1993 | |||
D63479.3 | - | 18 | 2180 | |||
D73409.1 | - | 17 | 2175 |
